missing translation for 'onlineSavingsMsg'
Learn More
Gibco™ Human PD-L1 (B7-H1) Fc Recombinant Protein, PeproTech® Código de producto.: 17872563

Gibco™ Human PD-L1 (B7-H1) Fc Recombinant Protein, PeproTech®

Código de producto. 17872563
500 μg, 500 microgramos
Click to view available options
Cantidad:
1 mg
100 μg
20 μg
250 μg
500 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
20 microgramos
250 microgramos
500 microgramos
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17872563

Marca: Gibco™ 31035500UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

PeproTech's CHO cell-derived Recombinant Human PD-L1 Fc is a glycosylated, disulfide-linked homodimer of 906 amino acid residues whose monomer consists of the 220-amino-acid length extracellular portion of PD-L1 fused to the 231-amino-acid length Fc portion of human IgG1 by two glycines. The calculated molecular weight of CHO cell-derived Recombinant Human PD-L1 Fc is 102.6 kDa, however, due to glycosylation, it migrates at an apparent molecular weight of approximately 160-170 kDa by SDS-PAGE analysis under non-reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Programmed death receptor ligand 1 (PD-L1, also called B7-H1) is a recently described B7 family member. To date, one specific receptor has been identified that can be ligated by PD-L1. This receptor, programmed death receptor 1 (PD-1), has been shown to negatively regulate T-cell receptor (TCR) signaling. Upon ligating its receptor, PD-L1 has been reported to decrease TCR-mediated proliferation and cytokine production. PD-L1 expression was found to be abundant on many murine and human cancers and could be further up-regulated upon IFN-gamma stimulation. Thus, PD-L1 might play an important role in tumor immune evasion.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso Q9NZQ7
Para utilizar con (aplicación) ELISA, Functional Assay, Western Blot
Formulación protein with no preservative
ID de gen (Entrez) 29126
Peso molecular 51.3 kDa
Nombre Human PD-L1 (B7-H1) Fc
Cantidad 500 μg
Fuente CHO cells
Estado normativo RUO
Concentración de endotoxinas <1 EU/ μg
Alias de gen A530045L16Rik; B7 homolog 1; B7-H; B7h1; B7-H1; Cd274; CD274 antigen; CD274 molecule; PDCD1 ligand 1; Pdcd1l1; Pdcd1lg1; Pdl1; PD-L1; pdl-1; Programmed cell death 1 ligand 1; programmed death ligand 1; RGD1566211; sCD274; soluble CD274; soluble PD L1; sPD L1
Nombre común PD-L1 (CD274)
Símbolo de gen Cd274
Actividad biológica Determined by its ability to induce adhesion in T-cell enriched PBMC cultures. The ED50 for this effect is 1.2-2.0 ug/ml
Conjugado Unconjugated
Recombinante Recombinant
Etiqueta de proteína Fc-tag
Secuencia FTVTVPKDLY VVEYGSNMTI ECKFPVEKQL DLAALIVYWE MEDKNIIQFV HGEEDLKVQH SSYRQRARLL KDQLSLGNAA LQITDVKLQD AGVYRCMISY GGADYKRITV KVNAPYNKIN QRILVVDPVT SEHELTCQAE GYPKAEVIWT SSDHQVLSGK TTTTNSKREE KLFNVTSTLR INTTTNEIFY CTFRRLDPEE NHTAELVIPE LPLAHPPNER GGPKSCDKTH TCPPCPAPEL LGGPSVFLFP PKPKDTLMIS RTPEVTCVVV DVSHEDPEVK FNWYVDGVEV HNAKTKPREE QYNSTYRVVS VLTVLHQDWL NGKEYKCKVS NKALPAPIEK TISKAKGQPR EPQVYTLPPS RDELTKNQVS LTCLVKGFYP SDIAVEWESN GQPENNYKTT PPVLDSDGSF FLYSKLTVDK SRWQQGNVFS CSVMHEALHN HYTQKSLSLS PGK
Contenido y almacenamiento -20°C
Sistema de expresión CHO cells
Formulario Lyophilized
Grado de pureza o calidad ≥ 95% by SDS-PAGE gel and HPLC analyses.
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
Gibco™ Human PD-L1 (B7-H1) Fc Recombinant Protein, PeproTech® >

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado