Learn More
Abnova™ Human PCDH20 Partial ORF (NP_073754, 395 a.a. - 494 a.a.) Recombinant Protein with GST-tag at N-terminal
Descripción
This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. This gene encodes a protein which contains 6 extracellular cadherin domains, a transmembrane domain and a cytoplasmic tail differing from those of the classical cadherins. Although its specific function is undetermined, the cadherin-related neuronal receptor is thought to play a role in the establishment and function of specific cell-cell connections in the brain. [provided by RefSeq]
Especificaciones
Especificaciones
Número de acceso | NP_073754 |
Para utilizar con (aplicación) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulación | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
ID de gen (Entrez) | 64881 |
Peso molecular | 36.74kDa |
Nombre | PCDH20 (Human) Recombinant Protein (Q01) |
Pruebas de control de calidad | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Cantidad | 10 μg |
Inmunógeno | RPPEIVPRYIANEIDGVVYLKELEPVNTPIAFFTIRDPEGKYKVNCYLDGEGPFRLSPYKPYNNEYLLETTKPMDYELQQFYEVAVVAWNSEGFHVKRVI |
Requisitos de almacenamiento | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Mostrar más |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.