missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human OSBPL10 Full-length ORF (NP_060254.2, 1 a.a. - 764 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Especificaciones
Número de acceso | NP_060254.2 |
---|---|
Para utilizar con (aplicación) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulación | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
ID de gen (Entrez) | 114884 |
Peso molecular | 110.4kDa |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
16197143
|
Abnova™
H00114884-P01.10UG |
10 ug |
Este artículo actualmente no está disponible o ha sido discontinuado. Ver la página del producto para posibles alternativas |
||||||
16107153
|
Abnova™
H00114884-P01.25UG |
25 ug |
Este artículo actualmente no está disponible o ha sido discontinuado. Ver la página del producto para posibles alternativas |
||||||
Descripción
This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Like most members, the encoded protein contains an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain. [provided by RefSeq]
Sequence: MERAVQGTDGGGGSNSSSRSSSRATSAGSSPSCSLAGRGVSSRSAAAGLGGGGSRSSPGSVAASPSGGGGRRREPALEGVLSKYTNLLQGWQNRYFVLDFEAGILQYFVNEQSKHQKPRGVLSLSGAIVSLSDEAPHMLVVYSANGEMFKLRAADAKEKQFWVTQLRACAKYHMEMNSKSAPSSRSRSLTLLPHGTPNSASPCSQRHLSVGAPGVVTITHHKSPAAARRAKSQYSGQLHEVREMMNQVEGQQKNLVHAIESLPGSGPLTALDQDLLLLKATSAATLSCLGECLNLLQQSVHQAGQPSQKPGASENILGWHGSKSHSTEQLKNGTLGSLPSASANITWAILPNSAEDEQTSQPEPEPNSGSELVLSEDEKSDNEDKEETELGVMEDQRSIILHLISQLKLGMDLTKVVLPTFILEKRSLLEMYADFMAHPDLLLAITAGATPEERVICFVEYYLTAFHEGRKGALAKKPYNPIIGETFHCSWEVPKDRVKPKRTASRSPASCHEHPMADDPSKSYKLRFVAEQVSHHPPISCFYCECEEKRLCVNTHVWTKSKFMGMSVGVSMIGEGVLRLLEHGEEYVFTLPSAYARSILTIPWVELGGKVSINCAKTGYSATVIFHTKPFYGGKVHRVTAEVKHNPTNTIVCKAHGEWNGTLEFTYNNGETKVIDTTTLPVYPKKIRPLEKQGPMESRNLWREVTRYLRLGDIDAATEQKRHLEEKQRVEERKRENLRTPWKPKYFIQEGDGWVYFNPLWKAHEspecificaciones
NP_060254.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
110.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
MERAVQGTDGGGGSNSSSRSSSRATSAGSSPSCSLAGRGVSSRSAAAGLGGGGSRSSPGSVAASPSGGGGRRREPALEGVLSKYTNLLQGWQNRYFVLDFEAGILQYFVNEQSKHQKPRGVLSLSGAIVSLSDEAPHMLVVYSANGEMFKLRAADAKEKQFWVTQLRACAKYHMEMNSKSAPSSRSRSLTLLPHGTPNSASPCSQRHLSVGAPGVVTITHHKSPAAARRAKSQYSGQLHEVREMMNQVEGQQKNLVHAIESLPGSGPLTALDQDLLLLKATSAATLSCLGECLNLLQQSVHQAGQPSQKPGASENILGWHGSKSHSTEQLKNGTLGSLPSASANITWAILPNSAEDEQTSQPEPEPNSGSELVLSEDEKSDNEDKEETELGVMEDQRSIILHLISQLKLGMDLTKVVLPTFILEKRSLLEMYADFMAHPDLLLAITAGATPEERVICFVEYYLTAFHEGRKGALAKKPYNPIIGETFHCSWEVPKDRVKPKRTASRSPASCHEHPMADDPSKSYKLRFVAEQVSHHPPISCFYCECEEKRLCVNTHVWTKSKFMGMSVGVSMIGEGVLRLLEHGEEYVFTLPSAYARSILTIPWVELGGKVSINCAKTGYSATVIFHTKPFYGGKVHRVTAEVKHNPTNTIVCKAHGEWNGTLEFTYNNGETKVIDTTTLPVYPKKIRPLEKQGPMESRNLWREVTRYLRLGDIDAATEQKRHLEEKQRVEERKRENLRTPWKPKYFIQEGDGWVYFNPLWKAH | |
RUO | |
OSBPL10 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
114884 | |
OSBPL10 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ20363/ORP10/OSBP9 | |
OSBPL10 | |
Recombinant | |
wheat germ expression system |