Learn More
Abnova™ Human OSBPL10 Full-length ORF (NP_060254.2, 1 a.a. - 764 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00114884-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Like most members, the encoded protein contains an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain. [provided by RefSeq]
Sequence: MERAVQGTDGGGGSNSSSRSSSRATSAGSSPSCSLAGRGVSSRSAAAGLGGGGSRSSPGSVAASPSGGGGRRREPALEGVLSKYTNLLQGWQNRYFVLDFEAGILQYFVNEQSKHQKPRGVLSLSGAIVSLSDEAPHMLVVYSANGEMFKLRAADAKEKQFWVTQLRACAKYHMEMNSKSAPSSRSRSLTLLPHGTPNSASPCSQRHLSVGAPGVVTITHHKSPAAARRAKSQYSGQLHEVREMMNQVEGQQKNLVHAIESLPGSGPLTALDQDLLLLKATSAATLSCLGECLNLLQQSVHQAGQPSQKPGASENILGWHGSKSHSTEQLKNGTLGSLPSASANITWAILPNSAEDEQTSQPEPEPNSGSELVLSEDEKSDNEDKEETELGVMEDQRSIILHLISQLKLGMDLTKVVLPTFILEKRSLLEMYADFMAHPDLLLAITAGATPEERVICFVEYYLTAFHEGRKGALAKKPYNPIIGETFHCSWEVPKDRVKPKRTASRSPASCHEHPMADDPSKSYKLRFVAEQVSHHPPISCFYCECEEKRLCVNTHVWTKSKFMGMSVGVSMIGEGVLRLLEHGEEYVFTLPSAYARSILTIPWVELGGKVSINCAKTGYSATVIFHTKPFYGGKVHRVTAEVKHNPTNTIVCKAHGEWNGTLEFTYNNGETKVIDTTTLPVYPKKIRPLEKQGPMESRNLWREVTRYLRLGDIDAATEQKRHLEEKQRVEERKRENLRTPWKPKYFIQEGDGWVYFNPLWKAHEspecificaciones
NP_060254.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
110.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ20363/ORP10/OSBP9 | |
OSBPL10 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
114884 | |
OSBPL10 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MERAVQGTDGGGGSNSSSRSSSRATSAGSSPSCSLAGRGVSSRSAAAGLGGGGSRSSPGSVAASPSGGGGRRREPALEGVLSKYTNLLQGWQNRYFVLDFEAGILQYFVNEQSKHQKPRGVLSLSGAIVSLSDEAPHMLVVYSANGEMFKLRAADAKEKQFWVTQLRACAKYHMEMNSKSAPSSRSRSLTLLPHGTPNSASPCSQRHLSVGAPGVVTITHHKSPAAARRAKSQYSGQLHEVREMMNQVEGQQKNLVHAIESLPGSGPLTALDQDLLLLKATSAATLSCLGECLNLLQQSVHQAGQPSQKPGASENILGWHGSKSHSTEQLKNGTLGSLPSASANITWAILPNSAEDEQTSQPEPEPNSGSELVLSEDEKSDNEDKEETELGVMEDQRSIILHLISQLKLGMDLTKVVLPTFILEKRSLLEMYADFMAHPDLLLAITAGATPEERVICFVEYYLTAFHEGRKGALAKKPYNPIIGETFHCSWEVPKDRVKPKRTASRSPASCHEHPMADDPSKSYKLRFVAEQVSHHPPISCFYCECEEKRLCVNTHVWTKSKFMGMSVGVSMIGEGVLRLLEHGEEYVFTLPSAYARSILTIPWVELGGKVSINCAKTGYSATVIFHTKPFYGGKVHRVTAEVKHNPTNTIVCKAHGEWNGTLEFTYNNGETKVIDTTTLPVYPKKIRPLEKQGPMESRNLWREVTRYLRLGDIDAATEQKRHLEEKQRVEERKRENLRTPWKPKYFIQEGDGWVYFNPLWKAH | |
RUO | |
OSBPL10 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |