missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ Human NR6A1 Partial ORF (NP_001480, 366 a.a. - 474 a.a.) Recombinant Protein with GST-tag at N-terminal Código de producto.: 16145381

Abnova™ Human NR6A1 Partial ORF (NP_001480, 366 a.a. - 474 a.a.) Recombinant Protein with GST-tag at N-terminal

Código de producto. 16145381
25 ug, 25 microgramos
Click to view available options
Cantidad:
10 ug
25 ug
Tamaño de la unidad:
10 microgramos
25 microgramos
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 16145381

Marca: Abnova™ H00002649Q01.25ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

Used for AP, Array, ELISA, WB-Re

This gene encodes an orphan nuclear receptor which is a member of the nuclear hormone receptor family. Its expression pattern suggests that it may be involved in neurogenesis and germ cell development. The protein can homodimerize and bind DNA, but in vivo targets have not been identified. The gene expresses at least alternatively spliced transcript variants. [provided by RefSeq]

Sequence: RLIYLYHKFHQLKVSNEEYACMKAINFLNQDIRGLTSASQLEQLNKRYWYICQDFTEYKYTHQPNRFPDLMMCLPEIRYIAGKMVNVPLEQLPLLFKVVLHSCKTSVGK

Specifications

Número de acceso NP_001480
Para utilizar con (aplicación) Antibody Production, ELISA, Protein Array, Western Blot
Formulación 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
ID de gen (Entrez) 2649
Peso molecular 37.73kDa
Nombre NR6A1 (Human) Recombinant Protein (Q01)
Pruebas de control de calidad 12.5% SDS-PAGE Stained with Coomassie Blue.
Cantidad 25 ug
Inmunógeno RLIYLYHKFHQLKVSNEEYACMKAINFLNQDIRGLTSASQLEQLNKRYWYICQDFTEYKYTHQPNRFPDLMMCLPEIRYIAGKMVNVPLEQLPLLFKVVLHSCKTSVGK
Requisitos de almacenamiento Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Estado normativo RUO
Alias de gen GCNF/GCNF1/NR61/RTR
Nombre común NR6A1
Símbolo de gen NR6A1
Especie Wheat Germ (in vitro)
Recombinante Recombinant
Etiqueta de proteína GST
Sistema de expresión wheat germ expression system
Formulario Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human NR6A1 Partial ORF (NP_001480, 366 a.a. - 474 a.a.) Recombinant Protein with GST-tag at N-terminal >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.