Learn More
Invitrogen™ Human NPTX2 (aa 168-245) Control Fragment Recombinant Protein
Recombinant Protein
Marca: Invitrogen™ RP108429
Descripción
Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-111466 (PA5-111466. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Neuronal pentraxin (NPTX) 2 was initially identified as a member of the pentraxin family having high homology to NPTX1. Unlike NPTX1 however, NPTX2 is expressed in testes, pancreas, liver, heart, skeletal muscle as well as brain. NPTX2 possesses the lectin properties common to the pentraxin family, promotes neurite outgrowth, and is rapidly regulated by neuronal activity. NPTX1 and NPTX2 form heterocomplexes that contribute to both activity-independent and -dependent excitatory synaptogenesis. Recently, NPTX2 has been found to be highly upregulated in Parkinsonian substantia nigra and localizes to Lewy bodies and Lewy neurites in sporadic Parkinson’s disease (PD), suggesting that it is involved in the pathway dysregulation that underlies PD.
Especificaciones
P47972 | |
Blocking Assay, Control | |
4885 | |
100 μL | |
apexin; Narp; neuro activity regulated petaxin; neuronal activity-regulated pentaxin; neuronal activity-regulated pentraxin; neuronal pentraxin 2; neuronal pentraxin II; Neuronal pentraxin-2; NP2; NP-II; Nptx2 | |
NPTX2 | |
Human | |
His-ABP-tag | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
≥5.0 mg/mL | |
1 M urea, PBS with no preservative; pH 7.4 | |
Human NPTX2 (aa 168-245) Control Fragment | |
RUO | |
NPTX2 | |
Unconjugated | |
Recombinant | |
ELERQLLRKVAELEDEKSLLHNETSAHRQKTESTLNALLQRVTELERGNSAFKSPDAFKVSLPLRTNYLYGKIKKTLP | |
E. coli | |
>80% by SDS-PAGE and Coomassie blue staining |
Certificados
Se requiere un número de lote para mostrar resultados para certificados. Para encontrar su número de lote en pedidos anteriores, utilice el área de estado de pedidos.
Número de lote | Tipo de certificado | Fecha | Catalog Number |
---|---|---|---|
79422643 | Certificado de análisis | 15/04/2025 |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.