Learn More
Abnova™ Human NF1 Partial ORF (NP_000258, 2719 a.a. - 2818 a.a.) Recombinant Protein with GST-tag at N-terminal
Descripción
This gene product appears to function as a negative regulator of the ras signal transduction pathway. Mutations in this gene have been linked to neurofibromatosis type 1, juvenile myelomonocytic leukemia and Watson syndrome. The mRNA for this gene is subject to RNA editing (CGA>UGA->Arg1306Term) resulting in premature translation termination. Alternatively spliced transcript variants encoding different isoforms have also been described for this gene. [provided by RefSeq]
Especificaciones
Especificaciones
Número de acceso | NP_000258 |
Para utilizar con (aplicación) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulación | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
ID de gen (Entrez) | 4763 |
Peso molecular | 36.74kDa |
Nombre | NF1 (Human) Recombinant Protein (Q01) |
Pruebas de control de calidad | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Cantidad | 25 ug |
Inmunógeno | DTYLPGIDEETSEESLLTPTSPYPPALQSQLSITANLNLSNSMTSLATSQHSPGIDKENVELSPTTGHCNSGRTRHGSASQVQKQRSAGSFKRNSIKKIV |
Requisitos de almacenamiento | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Mostrar más |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.