missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MOSC2 (aa 202-298) Control Fragment Recombinant Protein
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Descripción
Highest antigen sequence indentity to the following orthologs: Mouse (74%), Rat (74%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53276 (PA5-53276. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
As a component of the benzamidoxime prodrug-converting complex required to reduce N-hydroxylated prodrugs, such as benzamidoxime. Also able to reduce N(omega)-hydroxy-L-arginine (NOHA) and N(omega)-hydroxy-N(delta)-methyl-L-arginine (NHAM) into L-arginine and N(delta)-methyl-L-arginine, respectively.

Especificaciones
Especificaciones
Número de acceso | Q969Z3 |
Concentración | ≥5.0 mg/mL |
Para utilizar con (aplicación) | Blocking Assay, Control |
Formulación | 1 M urea, PBS with no preservative; pH 7.4 |
ID de gen (Entrez) | 54996 |
Nombre | Human MOSC2 (aa 202-298) Control Fragment |
Cantidad | 100 μL |
Estado normativo | RUO |
Alias de gen | 2810484M10Rik; mARC1; MARC2; Mg87; Mitochondrial amidoxime reducing component 2; moco sulfurase C-terminal domain-containing protein 2; MOCO sulphurase C-terminal domain containing 2; molybdenum cofactor sulfurase C-terminal domain-containing protein 2; MOSC domain-containing protein 2; MOSC domain-containing protein 2, mitochondrial; Mosc2; RP11-270A6.1 |
Nombre común | MOSC2 |
Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Invitrogen™ Human MOSC2 (aa 202-298) Control Fragment Recombinant Protein >
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido