Learn More
Invitrogen™ Human LRRC37A2 (aa 1614-1652) Control Fragment Recombinant Protein
Recombinant Protein
Marca: Invitrogen™ RP104444
Descripción
Highest antigen sequence indentity to the following orthologs: Mouse (38%), Rat (38%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61874 (PA5-61874. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Especificaciones
A6NM11 | |
Blocking Assay, Control | |
474170 | |
100 μL | |
c114 SLIT-like testicular protein; leucine rich repeat containing 37 member A2; leucine rich repeat containing 37, member A2; leucine-rich repeat containing 37 member A2; leucine-rich repeat-containing protein 37A2; LRRC37; LRRC37A2 | |
LRRC37A2 | |
Human | |
His-ABP-tag | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
≥5.0 mg/mL | |
1 M urea, PBS with no preservative; pH 7.4 | |
Human LRRC37A2 (aa 1614-1652) Control Fragment | |
RUO | |
LRRC37A2 | |
Unconjugated | |
Recombinant | |
DEEGFSRGIFRFLPWRGCSSRRESQDGLSSFGQPLWFKD | |
E. coli | |
>80% by SDS-PAGE and Coomassie blue staining |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.