missing translation for 'onlineSavingsMsg'
Learn More
Gibco™ Human Leptin Recombinant Protein, PeproTech® Código de producto.: 17832893

Gibco™ Human Leptin Recombinant Protein, PeproTech®

Código de producto. p-7245773
1 mg, 1 miligramo
Click to view available options
Cantidad:
1 mg
200 μg
5 mg
Tamaño de la unidad:
1 miligramo
200 microgramos
5 miligramos
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17832893

Marca: Gibco™ 300271MG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Recombinant Human Leptin is a 16.0 kDa protein containing 147 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Leptin is a 167 amino acid long protein hormone with important effects in regulating body weight, metabolism and reproductive function. Leptin is approximately 16 kDa in mass and encoded by the obese (ob) gene. Leptin is expressed predominantly by adipocytes, which fits with the idea that body weight is sensed as the total mass of fat in the body. Smaller amounts of leptin are also secreted by cells in the epithelium of the stomach and in the placenta. Leptin receptors are highly expressed in areas of the hypothalamus known to be important in regulating body weight, as well as in T lymphocytes and vascular endothelial cells. Leptin's effects on body weight are mediated through effects on hypothalamic centers that control feeding behavior and hunger, body temperature and energy expenditure. Leptin is involved in regulating food intake, energy expenditure, and adiposity through hypothalamic leptin receptors. Leptin promotes hematopoiesis, angiogenesis, wound healing, inflammation, immune responses, influences pubertal development and fetal growth. Studies have investigated the role of leptin in obesity, anorexia nervosa, insulin resistance, and hypertension. Leptin also has thermogenic actions and regulates enzymes of fatty acid oxidation. Severe hereditary obesity in rodents and humans can be caused by defects in leptin production.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P41159
Para utilizar con (aplicación) ELISA, Functional Assay, Western Blot
Formulación protein with no preservative
ID de gen (Entrez) 3952
Peso molecular 16 kDa
Nombre Human Leptin
Cantidad 1 mg
Fuente E. coli
Estado normativo RUO
Concentración de endotoxinas <1 EU/ μg
Alias de gen CD295 antigen; H-Leptin; HuB219; Lep; LEPD; LEP-R; Leptin; leptin (murine obesity homolog); leptin (obesity homolog, mouse); leptin precursor; Method: conceptual translation supplied by author.; OB; obese; obese protein; obese, mouse, homolog of; obesity; obesity factor; obesity protein; OB-R; OBS; truncated leptin
Nombre común Leptin
Símbolo de gen Lep
Actividad biológica PeproTech's Human Leptin is biologically active in the ob/ob mouse obesity model. The ob/ob mice were treated via intraperitoneal injection once daily at a dose of 5 ug Leptin/gm of body weight for 7 days. Significant effects on body weight, food consumption, and plasma glucose levels were observed compared to saline-treated controls.
Conjugado Unconjugated
Recombinante Recombinant
Secuencia MVPIQKVQDD TKTLIKTIVT RINDISHTQS VSSKQKVTGL DFIPGLHPIL TLSKMDQTLA VYQQILTSMP SRNVIQISND LENLRDLLHV LAFSKSCHLP WASGLETLDS LGGVLEASGY STEVVALSRL QGSLQDMLWQ LDLSPGC
Contenido y almacenamiento -20°C
Sistema de expresión E. coli
Formulario Lyophilized
Grado de pureza o calidad ≥ 98% by SDS-PAGE gel and HPLC analyses.
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
Gibco™ Human Leptin Recombinant Protein, PeproTech® >

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado