Learn More
Invitrogen™ Human Latexin (aa 1-106) Control Fragment Recombinant Protein
Recombinant Protein
Marca: Invitrogen™ RP89833
Descripción
Highest antigen sequence indentity to the following orthologs: Mouse (83%), Rat (83%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-82498 (PA5-82498. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene encodes the only known protein inhibitor of zinc-dependent metallocarboxypeptidases.
Especificaciones
Q9BS40 | |
Blocking Assay, Control | |
56925 | |
100 μL | |
ECI; endogenous carboxypeptidase inhibitor; Latexin; latexin protein; Lxn; MUM; Protein MUM; TCI; tissue carboxypeptidase inhibitor | |
LXN | |
Human | |
His-ABP-tag | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
≥5.0 mg/mL | |
1 M urea, PBS with no preservative; pH 7.4 | |
Human Latexin (aa 1-106) Control Fragment | |
RUO | |
Latexin | |
Unconjugated | |
Recombinant | |
MEIPPTNYPASRAALVAQNYINYQQGTPHRVFEVQKVKQASMEDIPGRGHKYHLKFAVEEIIQKQVKVNCTAEVLYPSTGQETAPEVNFTFEGETGKNPDEEDNTF | |
E. coli | |
>80% by SDS-PAGE and Coomassie blue staining |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.