Learn More
Invitrogen™ Human IMP4 (aa 111-194) Control Fragment Recombinant Protein
Recombinant Protein
Marca: Invitrogen™ RP107487
Descripción
Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66830 (PA5-66830. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
IMP4 forms a ternary complex with IMP3.
Especificaciones
Q96G21 | |
Blocking Assay, Control | |
92856 | |
100 μL | |
AA409888; AV031295; Brix domain-containing protein 4; BXDC4; D1Wsu40e; IMP4; IMP4 homolog, U3 small nucleolar ribonucleoprotein; IMP4, U3 small nucleolar ribonucleoprotein; IMP4, U3 small nucleolar ribonucleoprotein, homolog; IMP4, U3 small nucleolar ribonucleoprotein, homolog (yeast); LRRGT00017; RGD1308463; U3 small nucleolar ribonucleoprotein protein IMP4; U3 snoRNP protein 4 homolog; U3 snoRNP protein IMP4 | |
Imp4 | |
Human | |
His-ABP-tag | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
≥5.0 mg/mL | |
1 M urea, PBS with no preservative; pH 7.4 | |
Human IMP4 (aa 111-194) Control Fragment | |
RUO | |
IMP4 | |
Unconjugated | |
Recombinant | |
AQRMNRGRHEVGALVRACKANGVTDLLVVHEHRGTPVGLIVSHLPFGPTAYFTLCNVVMRHDIPDLGTMSEAKPHLITHGFSSR | |
E. coli | |
>80% by SDS-PAGE and Coomassie blue staining |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.