missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IL-29 (IFN lambda-1), AOF Recombinant Protein
E. coli Recombinant Protein
538.00€ - 3820.00€
Especificaciones
Número de acceso | Q8IU54 |
---|---|
Para utilizar con (aplicación) | Control, ELISA, Gel Shift, Western Blot |
Formulación | 0.1% TFA with no preservative |
ID de gen (Entrez) | 282618 |
Peso molecular | 19.9 kDa |
Descripción
IFN-lambdas constitute a novel family of interferons that mediate the induction of anti-viral protection in a wide variety of cells. There are three members in this family, namely lambda1, lambda2, and lambda3, which are also known as IL-29, IL-28A, and IL-28B, respectively. Similar to type I IFNs, IFN-lambdas activate an intracellular signaling to promote gene expression. IFN-lambdas mediate their anti-viral protection through a class II cytokine receptor complex composed of two essential receptor proteins, CRF2-12/IFN-lambdaR1, which is unique to IFN-lambdas, and CFR2-4/IL-10R2, which is shared with IL-10, IL-22, and IL-26 receptors. The IFN-lambdas, IL-28 and IL-29, have recently been reported to prime dendritic cells to induce proliferation of Foxp3-bearing regulatory T cells. DCs that differentiate in response to IFN-lambdas express high levels of class I and II MHC gene products, but low levels of costimulatory molecules. Moreover, these cells can specifically induce IL-2-dependent proliferation of CD4+CD25+FOXP3+ T cells with contact-dependent suppressive activity.- Purity: >95% by SDS-Page
- Endotoxin Concentration: AX41 ≤EUs/μg
- Protein Length: 179 aa.
- Molecular Weight: 19.9 kDa
- Animal Origin Free: Yes
Especificaciones
Q8IU54 | |
0.1% TFA with no preservative | |
19.9 kDa | |
E. coli | |
RUO | |
IFNL1 | |
Unconjugated | |
Lyophilized |
Control, ELISA, Gel Shift, Western Blot | |
282618 | |
Human IL-29, AOF | |
-20°C | |
Cytokine Zcyto21; FNL1; IFN lambda1; IFNL1; IFN-lambda-1; IL29; IL-29; ILN; interferon lambda 1; Interferon lambda1; Interferon lambda-1; interferon, lambda 1; interferon-lambda-1; Interleukin; interleukin 29 (interferon, lambda 1); Interleukin-29; ZCYTO21 | |
Human | |
Recombinant | |
>95% |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Invitrogen™ Human IL-29 (IFN lambda-1), AOF Recombinant Protein