Learn More
Invitrogen™ Human FLII (aa 776-904) Control Fragment Recombinant Protein
Recombinant Protein
Marca: Invitrogen™ RP89498
Descripción
Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-82369 (PA5-82369. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Mahogunin (MGRN1) is a C3HC4 RING-containing protein with E3 ubiquitin ligase activity in vitro.
Especificaciones
Q13045 | |
Blocking Assay, Control | |
2314 | |
100 μL | |
3632430F08Rik; FLI; Fli1; flightless I actin binding protein; flightless I homolog; flightless I homolog (Drosophila); Flii; FLII actin remodeling protein; FLII, actin remodeling protein; Fliih; FLIL; Protein flightless-1 homolog | |
FLII | |
Human | |
His-ABP-tag | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
≥5.0 mg/mL | |
1 M urea, PBS with no preservative; pH 7.4 | |
Human FLII (aa 776-904) Control Fragment | |
RUO | |
FLII | |
Unconjugated | |
Recombinant | |
YILDCWSDVFIWLGRKSPRLVRAAALKLGQELCGMLHRPRHATVSRSLEGTEAQVFKAKFKNWDDVLTVDYTRNAEAVLQSPGLSGKVKRDAEKKDQMKADLTALFLPRQPPMSLAEAEQLMEEWNEDL | |
E. coli | |
>80% by SDS-PAGE and Coomassie blue staining |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.