Learn More
Invitrogen™ Human ECM2 (aa 281-366) Control Fragment Recombinant Protein
Descripción
Highest antigen sequence indentity to the following orthologs: Mouse (69%), Rat (69%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57239 (PA5-57239. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Especificaciones
Especificaciones
Número de acceso | O94769 |
Concentración | ≥5.0 mg/mL |
Para utilizar con (aplicación) | Blocking Assay, Control |
Formulación | 1 M urea, PBS with no preservative; pH 7.4 |
ID de gen (Entrez) | 1842 |
Nombre | Human ECM2 (aa 281-366) Control Fragment |
Cantidad | 100 μL |
Estado normativo | RUO |
Alias de gen | 9030618O22Rik; BC065151; ECM2; Extracellular matrix protein 2; extracellular matrix protein 2, female organ and adipocyte specific; extracellular matrix protein 2-like; LOC100361383; Matrix glycoprotein SC1/ECM2; Tenonectin |
Nombre común | ECM2 |
Mostrar más |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.