missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DGKG Full-length ORF (NP_001074213.1, 1 a.a. - 766 a.a.) Recombinant Protein with GST-tag at N-terminal

Código de producto. 16153891
25 ug, 25 microgramos
Click to view available options
Cantidad:
10 ug
25 ug
Tamaño de la unidad:
10 microgramos
25 microgramos
Este artículo no se puede devolver. Vea la política de devoluciones
Este artículo no se puede devolver. Vea la política de devoluciones

Used for AP, Array, ELISA, WB-Re

This gene encodes an enzyme that is a member of the type I subfamily of diacylglycerol kinases, which are involved in lipid metabolism. These enzymes generate phosphatidic acid by catalyzing the phosphorylation of diacylglycerol, a fundamental lipid second messenger that activates numerous proteins, including protein kinase C isoforms, Ras guanyl nucleotide-releasing proteins and some transient receptor potential channels. Diacylglycerol kinase gamma has been implicated in cell cycle regulation and in the negative regulation of macrophage differentiation in leukemia cells. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MGEERWVSLTPEEFDQLQKYSEYSSKKIKDALTEFNEGGSLKQYDPHEPISYDVFKLFMRAYLEVDLPQPLSTHLFLAFSQKPRHETSDHPTEGASNSEANSADTNIQNADNATKADEACAPDTESNMAEKQAPAEDQVAATPLEPPVPRSSSSESPVVYLKDVVCYLSLLETGRPQDKLEFMFRLYDSDENGLLDQAEMDCIVNQMLHIAQYLEWDPTELRPILKEMLQGMDYDRDGFVSLQEWVHGGMTTIPLLVLLGMDDSGSKGDGRHAWTMKHFKKPTYCNFCHIMLMGVRKQGLCCTYCKYTVHERCVSRNIPGCVKTYSKAKRSGEVMQHAWVEGNSSVKCDRCHKSIKCYQSVTARHCVWCRMTFHRKCELSTLCDGGELRDHILLPTSICPITRDRPGEKSDGCVSAKGELVMQYKIIPTPGTHPLLVLVNPKSGGRQGERLNFFRDTPDFRVLACGGDGTVGWILDCIDKANFAKHPPVAVLPLGTGNDLARCLRWGGGYEGGSLTKILKDIEQSPLVMLDRWHLEVIPREEVENGDQVPYSIMNNYFSIGVDASIAHRFHVMREKHPEKFNSRMKNKLWYFEFGTSETFAATCKKLHDHIELECDGVGVDLSNIFLEGIAILNIPSMYGGTNLWGENKKNRAVIRESRKGVTDPKELKFCVQDLSDQLLEVVGLEGAMEMGQIYTGLKSAGRRLAQCASVTIRTNKLLPMQVDGEPWMQPCCTIKITHKNQAPMMMGPPQKSSFFSLRRKSRSKD

Especificaciones

Número de acceso NP_001074213.1
Para utilizar con (aplicación) Antibody Production, Protein Array, ELISA, Western Blot
Formulación 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
ID de gen (Entrez) 1608
Peso molecular 112.7kDa
Nombre DGKG (Human) Recombinant Protein (P01)
Método de purificación Glutathione Sepharose 4 Fast Flow
Pruebas de control de calidad 12.5% SDS-PAGE Stained with Coomassie Blue.
Cantidad 25 ug
Inmunógeno MGEERWVSLTPEEFDQLQKYSEYSSKKIKDALTEFNEGGSLKQYDPHEPISYDVFKLFMRAYLEVDLPQPLSTHLFLAFSQKPRHETSDHPTEGASNSEANSADTNIQNADNATKADEACAPDTESNMAEKQAPAEDQVAATPLEPPVPRSSSSESPVVYLKDVVCYLSLLETGRPQDKLEFMFRLYDSDENGLLDQAEMDCIVNQMLHIAQYLEWDPTELRPILKEMLQGMDYDRDGFVSLQEWVHGGMTTIPLLVLLGMDDSGSKGDGRHAWTMKHFKKPTYCNFCHIMLMGVRKQGLCCTYCKYTVHERCVSRNIPGCVKTYSKAKRSGEVMQHAWVEGNSSVKCDRCHKSIKCYQSVTARHCVWCRMTFHRKCELSTLCDGGELRDHILLPTSICPITRDRPGEKSDGCVSAKGELVMQYKIIPTPGTHPLLVLVNPKSGGRQGERLNFFRDTPDFRVLACGGDGTVGWILDCIDKANFAKHPPVAVLPLGTGNDLARCLRWGGGYEGGSLTKILKDIEQSPLVMLDRWHLEVIPREEVENGDQVPYSIMNNYFSIGVDASIAHRFHVMREKHPEKFNSRMKNKLWYFEFGTSETFAATCKKLHDHIELECDGVGVDLSNIFLEGIAILNIPSMYGGTNLWGENKKNRAVIRESRKGVTDPKELKFCVQDLSDQLLEVVGLEGAMEMGQIYTGLKSAGRRLAQCASVTIRTNKLLPMQVDGEPWMQPCCTIKITHKNQAPMMMGPPQKSSFFSLRRKSRSKD
Requisitos de almacenamiento Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Estado normativo RUO
Alias de gen DAGK3/DGK-GAMMA/MGC104993/MGC133330
Nombre común DGKG
Símbolo de gen DGKG
Especie Wheat Germ (in vitro)
Recombinante Recombinant
Etiqueta de proteína GST
Sistema de expresión wheat germ expression system
Formulario Liquid
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
Abnova™ Human DGKG Full-length ORF (NP_001074213.1, 1 a.a. - 766 a.a.) Recombinant Protein with GST-tag at N-terminal >

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado