Learn More
Abnova™ Human COL4A3BP Full-length ORF (NP_112729.1, 1 a.a. - 598 a.a.) Recombinant Protein with GST-tag at N-terminal
Descripción
This gene encodes a kinase that specifically phosphorylates the N-terminal region of the non-collagenous domain of the alpha 3 chain of type IV collagen, known as the Goodpasture antigen. Goodpasture disease is the result of an autoimmune response directed at this antigen. One isoform of this protein is also involved in ceramide intracellular transport. Three transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Especificaciones
Especificaciones
Número de acceso | NP_112729.1 |
Para utilizar con (aplicación) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulación | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
ID de gen (Entrez) | 10087 |
Peso molecular | 94.4kDa |
Nombre | COL4A3BP (Human) Recombinant Protein (P01) |
Método de purificación | Glutathione Sepharose 4 Fast Flow |
Pruebas de control de calidad | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Cantidad | 10 μg |
Inmunógeno | MSDNQSWNSSGSEEDPETESGPPVERCGVLSKWTNYIHGWQDRWVVLKNNALSYYKSEDETEYGCRGSICLSKAVITPHDFDECRFDISVNDSVWYLRAQDPDHRQQWIDAIEQHKTESGYGSESSLRRHGSMVSLVSGASGYSATSTSSFKKGHSLREKLAEMETFRDILCRQVDTLQKYFDACADAVSKDELQRDKVVEDDEDDFPTTRSDGDFLHSTNGNKEKLFPHVTPKGINGIDFKGEAITFKATTAGILATLSHCIELMVKREDSWQKRLDKETEKKRRTEEAYKNAMTELKKKSHFGGPDYEEGPNSLINEEEFFDAVEAALDRQDKIEEQSQSEKVRLHWPTSLPSGDAFSSVGTHRFVQKVEEMVQNHMTYSLQDVGGDANWQLVVEEGEMKVYRREVEENGIVLDPLKATHAVKGVTGHEVCNYFWNVDVRNDWETTIENFHVVETLADNAIIIYQTHKRVWPASQRDVLYLSVIRKIPALTENDPETWIVCNFSVDHDSAPLNNRCVRAKINVAMICQTLVSPPEGNQEISRDNILCKITYVANVNPGGWAPASVLRAVAKREYPKFLKRFTSYVQEKTAGKPILF |
Mostrar más |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.