Learn More
Abnova™ Human CNN3 Partial ORF (NP_001830, 230 a.a. - 329 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00001266-Q01.10ug
Descripción
This gene encodes a protein with a markedly acidic C terminus; the basic N-terminus is highly homologous to the N-terminus of a related gene, CNN1. Members of the CNN gene family all contain similar tandemly repeated motifs. This encoded protein is associated with the cytoskeleton but is not involved in contraction. [provided by RefSeq]
Sequence: DQKLTLQPVDNSTISLQMGTNKVASQKGMSVYGLGRQVYDPKYCAAPTEPVIHNGSQGTGTNGSEISDSDYQAEYPDEYHGEYQDDYPRDYQYSDQGIDYEspecificaciones
NP_001830 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DQKLTLQPVDNSTISLQMGTNKVASQKGMSVYGLGRQVYDPKYCAAPTEPVIHNGSQGTGTNGSEISDSDYQAEYPDEYHGEYQDDYPRDYQYSDQGIDY | |
RUO | |
CNN3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
1266 | |
CNN3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CNN3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.