Learn More
Invitrogen™ Human CEBPZOS (aa 32-72) Control Fragment Recombinant Protein
Recombinant Protein
Marca: Invitrogen™ RP99713
Descripción
Highest antigen sequence indentity to the following orthologs: Mouse (80%), Rat (80%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84174 (PA5-84174. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
CEBPZOS is a protein coding gene. Gene ontology (GO) annotation include integral component of membrane; mitochondrial membrane.
Especificaciones
A8MTT3 | |
Blocking Assay, Control | |
100505876 | |
100 μL | |
CEBPZ antisense RNA 1; CEBPZ opposite strand; CEBPZ-AS1; CEBPZOS; Protein CEBPZOS | |
CEBPZOS | |
Human | |
His-ABP-tag | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
≥5.0 mg/mL | |
1 M urea, PBS with no preservative; pH 7.4 | |
Human CEBPZOS (aa 32-72) Control Fragment | |
RUO | |
CEBPZOS | |
Unconjugated | |
Recombinant | |
SKMHTSQDFRQTMSKKYPFILEVYYKSTEKSGMYGIRELDQ | |
E. coli | |
>80% by SDS-PAGE and Coomassie blue staining |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.