Learn More
Abnova™ Human CDH4 Partial ORF (NP_001785, 635 a.a. - 734 a.a.) Recombinant Protein with GST-tag at N-terminal
Descripción
This gene is a classical cadherin from the cadherin superfamily. The encoded protein is a calcium-dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Based on studies in chicken and mouse, this cadherin is thought to play an important role during brain segmentation and neuronal outgrowth. In addition, a role in kidney and muscle development is indicated. Of particular interest are studies showing stable cis-heterodimers of cadherins 2 and 4 in cotransfected cell lines. Previously thought to interact in an exclusively homophilic manner, this is the first evidence of cadherin heterodimerization. [provided by RefSeq]
Especificaciones
Especificaciones
Número de acceso | NP_001785 |
Para utilizar con (aplicación) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulación | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
ID de gen (Entrez) | 1002 |
Peso molecular | 36.74kDa |
Nombre | CDH4 (Human) Recombinant Protein (Q01) |
Pruebas de control de calidad | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Cantidad | 25 ug |
Inmunógeno | AADADVDPNIGPYVFELPFVPAAVRKNWTITRLNGDYAQLSLRILYLEAGMYDVPIIVTDSGNPPLSNTSIIKVKVCPCDDNGDCTTIGAVAAAGLGTGA |
Requisitos de almacenamiento | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Mostrar más |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.