Learn More
Invitrogen™ Human CD3 delta Fc Chimera Flag-tag Recombinant Protein
Recombinant Protein
Marca: Invitrogen™ RP88000
371.70 EUR hasta el 2025-06-30
Use el código promocional "24108" para aplicar la oferta.
Descripción
It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (1000 μL) be added to the vial to prepare a stock solution of 0.25 mg/mL. Concentration is measured by UV.
CD3d is part of the T-cell receptor/CD3 complex (TCR/CD3 complex) and is involved in T-cell development and signal transduction. The CD3d protein represents the delta subunit of the CD3 complex, and along with four other CD3 subunits, binds either TCR alpha/beta or TCR gamma/delta to form the TCR/CD3 complex on the surface of T-cells. Defects in the CD3d gene are a cause of severe combined immunodeficiency autosomal recessive T-cell-negative/B-cell-positive/NK-cell-positive (SCIDBNK). Two transcript variants encoding different isoforms of CD3d have been found. Other variants of CD3d may also exist, but the full-length nature of their transcripts have yet to be defined.
Especificaciones
P04234 | |
Control | |
PBS with 0.01% Tween 80, 5% mannitol, 5% trehalose and no preservative; pH 7.4 | |
47-51 kDa | |
SDS-PAGE | |
RUO | |
CD3 antigen delta polypeptide; CD3 antigen, delta polypeptide; CD3 antigen, delta subunit; CD3 delta; CD3 molecule delta polypeptide; CD3 molecule, delta; CD3D; CD3d antigen, delta polypeptide (TiT3 complex); CD3d molecule; CD3d molecule, delta (CD3-TCR complex); CD3-DELTA; IMD19; OKT3, delta chain; T3D; T-cell receptor T3 delta chain; T-cell surface glycoprotein CD3 delta chain | |
Cd3d | |
Unconjugated | |
DYKDDDDK-tag, Fc-tag | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
0.25 mg/mL | |
Lyophilized | |
915 | |
Human CD3 delta Fc Chimera Flag-tag | |
100 μg | |
<1.0 EU/μg | |
CD3d | |
Protein | |
Recombinant | |
Human CD3 delta, amino acids Met1-Ala105 (Accession # NP_000723.1) with a C-terminal Flag-tagged human IgG1 Fc region | |
HEK293 cells | |
>85% as determined by SDS-PAGE. |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.