Learn More
Invitrogen™ Human C7orf49 (aa 48-154) Control Fragment Recombinant Protein
Descripción
Highest antigen sequence indentity to the following orthologs: Mouse (70%), Rat (70%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54132 (PA5-54132. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Especificaciones
Especificaciones
Número de acceso | Q9BWK5 |
Concentración | ≥5.0 mg/mL |
Para utilizar con (aplicación) | Blocking Assay, Control |
Formulación | 1 M urea, PBS with no preservative; pH 7.4 |
ID de gen (Entrez) | 78996 |
Nombre | Human C7orf49 (aa 48-154) Control Fragment |
Cantidad | 100 μL |
Estado normativo | RUO |
Alias de gen | C7orf49; Cell cycle regulator of NHEJ; Cell cycle regulator of non-homologous end joining; chromosome 7 open reading frame 49; CYREN; Modulator of retrovirus infection homolog; MRI |
Nombre común | C7orf49 |
Mostrar más |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.