Learn More
Invitrogen™ Human C17orf50 (aa 9-85) Control Fragment Recombinant Protein
Recombinant Protein
Marca: Invitrogen™ RP106037
Descripción
Highest antigen sequence indentity to the following orthologs: Mouse (52%), Rat (52%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65351 (PA5-65351. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
C17orf50 is a protein coding gene.
Especificaciones
Q8WW18 | |
Blocking Assay, Control | |
146853 | |
100 μL | |
C17orf50; chromosome 17 open reading frame 50; uncharacterized protein C17orf50 | |
C17orf50 | |
Human | |
His-ABP-tag | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
≥5.0 mg/mL | |
1 M urea, PBS with no preservative; pH 7.4 | |
Human C17orf50 (aa 9-85) Control Fragment | |
RUO | |
C17orf50 | |
Unconjugated | |
Recombinant | |
PLWKKETEELRAEDAEQEEGKEGSEDEDEDNQRPLEDSATEGEEPPRVAEEGEGRERRSVSYCPLRQESSTQQVALL | |
E. coli | |
>80% by SDS-PAGE and Coomassie blue staining |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.