missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ATP7A (aa 1102-1223) Control Fragment Recombinant Protein
missing translation for 'orderingAttributeHoverText'
Cantidad:
100 μL
missing translation for 'unitSize'
100 microlitros
Descripción
Highest antigen sequence indentity to the following orthologs: Mouse (85%), Rat (85%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53003 (PA5-53003. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
ATP7A (also known as Copper-transporting ATPase 1) functions as a transmembrane copper-translocating P-type ATPase and plays a vital role in systemic copper absorption in the gut and copper reabsorption in the kidney. Polarized epithelial cells such as Madin-Darby canine kidney cells are a physiologically relevant model for systemic copper absorption and reabsorption in vivo. Although ATP7A is not detectable in most normal tissues, it is expressed in a considerable fraction of many common tumor types. Increased expression of ATP7A renders cells resistant to cisplatin and carboplatin. Mutations in the ATP7A gene result in Menkes disease, which is fatal in early childhood.

Especificaciones
Especificaciones
Número de acceso | Q04656 |
Concentración | ≥5.0 mg/mL |
Para utilizar con (aplicación) | Blocking Assay, Control |
Formulación | 1 M urea, PBS with no preservative; pH 7.4 |
ID de gen (Entrez) | 538 |
Nombre | Human ATP7A (aa 1102-1223) Control Fragment |
Cantidad | 100 μL |
Estado normativo | RUO |
Alias de gen | ATP 7 A; ATP7A; ATPase copper transporting alpha; ATPase, Cu++ transporting, alpha polypeptide; copper pump 1; Copper-transporting ATPase 1; Cu++-transporting P-type ATPase; DSMAX; FLJ17790; MC 1; MC1; Menke; Menkes disease-associated protein; menkes disease-associated protein homolog; Menkes protein; Menkes syndrome; MK; MNK; OHS; OTTHUMP00000062077; SMA x 3 |
Nombre común | ATP7A |
Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Invitrogen™ Human ATP7A (aa 1102-1223) Control Fragment Recombinant Protein >
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido