Learn More
Abnova™ Human ABL1 Partial ORF (NP_005148.2, 931 a.a. - 1030 a.a.) Recombinant Protein with GST-tag at N-terminal
Descripción
The ABL1 protooncogene encodes a cytoplasmic and nuclear protein tyrosine kinase that has been implicated in processes of cell differentiation, cell division, cell adhesion, and stress response. Activity of c-Abl protein is negatively regulated by its SH3 domain, and deletion of the SH3 domain turns ABL1 into an oncogene. The t(9;22) translocation results in the head-to-tail fusion of the BCR (MIM:151410) and ABL1 genes present in many cases of chronic myelogeneous leukemia. The DNA-binding activity of the ubiquitously expressed ABL1 tyrosine kinase is regulated by CDC2-mediated phosphorylation, suggesting a cell cycle function for ABL1. The ABL1 gene is expressed as either a 6- or 7-kb mRNA transcript, with alternatively spliced first exons spliced to the common exons 2-11. [provided by RefSeq]
Especificaciones
Especificaciones
Número de acceso | NP_005148.2 |
Para utilizar con (aplicación) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulación | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
ID de gen (Entrez) | 25 |
Peso molecular | 36.74kDa |
Nombre | ABL1 (Human) Recombinant Protein (Q01) |
Método de purificación | Glutathione Sepharose 4 Fast Flow |
Pruebas de control de calidad | 12.5% SDS-PAGE Stained with Coomassie Blue |
Cantidad | 2 μg |
Inmunógeno | KTKATSLVDAVNSDAAKPSQPGEGLKKPVLPATPKPQSAKPSGTPISPAPVPSTLPSASSALAGDQPSSTAFIPLISTRVSLRKTRQPPERIASGAITKG |
Mostrar más |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.