missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ HMGN2 Recombinant Protein

Marca:  Abnova™ H00003151-P01.25ug

 Ver más versiones de este producto

Código de producto. 16166231

  • 521.00€ / 25 microgramos

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Especificaciones

Especificaciones

AAH14644
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
35.6
8
Glutathione Sepharose 4 Fast Flow
25 μg
MPKRKAEEDAKGDKAKVKDEPQRRSARLSAKPAPPKPEPKPKKAPAKKGEKVPKGKKGKADAGKEGNNPAENGDAKTDQAQKAEGAGDAK
HMG17/MGC5629/MGC88718
HMGN2
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
3151
HMGN2 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HMGN2
Human
Recombinant
Solution
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

One moment while we fetch your results.
Certificados
Promociones

Promociones

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Product Title
Abnova™ HMGN2 Recombinant Protein > 25μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.