missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ HLA-DPB1 (Human) Recombinant Protein

Human HLA-DPB1 full-length ORF ( AAH07963, 30 a.a. - 258 a.a.) recombinant protein with GST-tag at N-terminal.

Marca:  Abnova™ H00003115-P02.25ug

 Ver más versiones de este producto

Código de producto. 16176151

  • 521.00€ / 25 microgramos

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

  • Sequence: RATPENYLFQGRQECYAFNGTQRFLERYIYNREEFVRFDSDVGEFRAVTELGRPAAEYWNSQKDILEEKRAVPDRMCRHNYELGGPMTLQRRVQPRVNVSPSKKGPLQHHNLLVCHVTDFYPGSIQVRWFLNGQEETAGVVSTNLIRNGDWTFQILVMLEMTPQQGDVYTCQVEHTSLDSPVTVEWKAQSDSARSKTLTGAGGFVLGLIICGVGIFMHRRSKKVQRGSA
Especificaciones

Especificaciones

AAH07963
major histocompatibility complex, class II, DP beta 1
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HLA-DPB1
GST
3115
Wheat germ expression system
25 μg
DPB1, HLA-DP1B
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Promociones

Promociones

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ HLA-DPB1 (Human) Recombinant Protein > Quantity: 25 μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.