missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRP94 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14700-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody was generated against the C-terminal region of full-length HSP90b1.
GRP94 is a 803 amino acid protein belonging to the heat shock protein 90 family. It acts as a molecular chaperone that functions in the processing and transport of secreted proteins. GRP94 and its N-terminal fragment stimulates CTL expansion and maturation of human monocyte-derived dendritic cells (MDDC). It plays a central role in innate as well as acquired immunity, maturation and chemotaxis of dendritic cells, Ab production, cross-priming, as a potential marker in breast cancer and is a peptide acceptor in endoplasmic reticulum and an accessory to peptide loading of MHC class I molecules. Expression of GRP94 suppressed A23187-induced apoptosis and stabilized calcium homeostasis. GRP94 is expressed in melanoma or liver metastases of colon carcinoma cells, human gastric carcinoma BGC-823 cells.Especificaciones
GRP94 | |
Polyclonal | |
Unconjugated | |
HSP90B1 | |
ECGP, Endoplasmin, GP96, gp96 homolog, GRP 94, GRP94, HSP90B1, TRA1, Tumor rejection antigen 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22027, 362862, 7184 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P08113, P14625, Q66HD0 | |
Hsp90b1 | |
GRP94 Fusion Protein Ag6344 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
GRP94 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido