Learn More
GPR65 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20306-1-AP-150UL
Description
The GPR65 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human GPR65. This antibody recognizes human antigen. The GPR65 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
GPR65 | |
Polyclonal | |
Unconjugated | |
150 μL | |
8477 | |
IgG |
Immunohistochemistry, ELISA | |
393 μg/mL | |
Rabbit | |
Primary | |
Human |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.