missing translation for 'onlineSavingsMsg'
Learn More

GPR65 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Código de producto. p-7223487 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Unit Size:
150 microlitros
20 microlitros
Product Code. Cantidad unitSize
16819834 20 μL 20 microlitros
16809834 150 μL 150 microlitros
2 options
This item is not returnable. View return policy

Product Code. 16819834

Brand: Proteintech 203061AP20UL

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The GPR65 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human GPR65. This antibody recognizes human antigen. The GPR65 antibody has been validated for the following applications: IHC, ELISA analysis.

Specifications

Antígeno GPR65
Aplicaciones Immunohistochemistry, ELISA
Clasificación Polyclonal
Concentración 393 μg/mL
Conjugado Unconjugated
Especie del huésped Rabbit
Cantidad 20 μL
Primario o secundario Primary
ID de gen (Entrez) 8477
Especies diana Human
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.