Learn More
Proteintech FlexAble CoraLite Plus 555 Antibody Labeling Kit for Human IgG
FlexAble CoraLite Plus 555 Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite Plus 555 in any buffer condition to Human IgG primary antibodies from any supplier.
90.00€ - 889.00€
Especificaciones
Formato Liquid
Ver más especificaciones
Productos 3
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
17981427
Ver documentos
Proteintech
KFA105-10
10 Tests
90.00€
paquete de 10
18084735
Ver documentos
Proteintech
KFA105-200
100 Tests
889.00€
200 reacciones
18044235
Ver documentos
Proteintech
KFA105-50
50 Tests
419.00€
50 reacciones
Descripción
Descripción
FlexAble CoraLite™ Plus 555 Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite™ Plus 555 in any buffer condition to Human IgG primary antibodies from any supplier. One labeling reaction requires only 0.5 μg of antibody - regardless of the antibody concentration. Label your Human IgG antibody in<10 min without any additional equipment.
Especificaciones
Especificaciones
Liquid
Documentos
Product Certifications
Título del producto
Proteintech FlexAble CoraLite Plus 555 Antibody Labeling Kit for Human IgG