Learn More
Abnova™ ENO1 Recombinant Protein
Human ENO1 full-length ORF recombinant protein with GST-tag at N-terminal
344.00€ - 521.00€
Especificaciones
Número de acceso | AAH11130.1 |
---|---|
Para utilizar con (aplicación) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulación | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
ID de gen (Entrez) | 2023 |
Peso molecular | 73.48 |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
16164531
|
Abnova™
H00002023-P01.10ug |
10 μg |
344.00€
10 microgramos |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
16174531
|
Abnova™
H00002023-P01.25ug |
25 μg |
521.00€
25 microgramos |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
This gene encodes one of three enolase isoenzymes found in mammals; it encodes alpha-enolase, a homodimeric soluble enzyme, and also encodes a shorter monomeric structural lens protein, tau-crystallin. The two proteins are made from the same message. The full length protein, the isoenzyme, is found in the cytoplasm. The shorter protein is produced from an alternative translation start, is localized to the nucleus, and has been found to bind to an element in the c-myc promoter. A pseudogene has been identified that is located on the other arm of the same chromosome.
- Molecular Weight: 73.48kDa
- Preparation Method: in vitro wheat germ expression system
- Purification: Glutathione Sepharose 4 Fast Flow
- Storage Buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH 8.0 in the elution buffer
Sequence: MSILKIHAREIFDSRGNPTVEVDLFTSKGLFRAAVPSGASTGIYEALELRDNDKTRYMGKGVSKAVEHINKTIAPALVSKKLNVTEQEKIDKLMIEMDGTENKSKFGANAILGVSLAVCKAGAVEKGVPLYRHIADLAGNSEVILPVPAFNVINGGSHAGNKLAMQEFMILPVGAANFREAMRIGAEVYHNL
KNVIKEKYGKDATNVGDEGGFAPNILENKEGLELLKTAIGKAGYTDKVVIGMDVAASEFFRSGKYDLDFKSPDDPSRYISPDQLADLYKSFIKDYPVVSIEDPFDQDDWGAWQKFTASAGIQVVGDDLTVTNPKRIAKAVNEKSCNCLLLKVNQIGSVTESLQACKLAQANGWGVMVSHRSGETEDTFIADLVVGLCTGQIKTGAPCRSERLAKYNQLLRIEEGLGSKAKFAGRNFRNPLAK
Best when used within three months from the date of receipt.
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Especificaciones
AAH11130.1 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
73.48 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ENO1 | |
Human | |
Recombinant | |
Solution |
Antibody Production, Protein Array, ELISA, Western Blot | |
2023 | |
ENO1 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSILKIHAREIFDSRGNPTVEVDLFTSKGLFRAAVPSGASTGIYEALELRDNDKTRYMGKGVSKAVEHINKTIAPALVSKKLNVTEQEKIDKLMIEMDGTENKSKFGANAILGVSLAVCKAGAVEKGVPLYRHIADLAGNSEVILPVPAFNVINGGSHAGNKLAMQEFMILPVGAANFREAMRIGAEVYHNLKNVIKEKYGKDATNVGDEGGFAPNILENKEGLELLKTAIGKAGYTDKVVIGMDVAASEFFRSGKYDLDFKSPDDPSRYISPDQLADLYKSFIKDYPVVSIEDPFDQDDWGAWQKFTASAGIQVVGDDLTVTNPKRIAKAVNEKSCNCLLLKVNQIGSVTESLQACKLAQANGWGVMVSHRSGETEDTFIADLVVGLCTGQIKTGAPCRSERLAKYNQLLRIEEGLGSKAKFAGRNFRNPLAK | |
ENO1L1/MBP-1/MPB1/NNE/PPH | |
ENO1 | |
Wheat Germ (in vitro) | |
GST |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.