missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E-selectin Recombinant Rabbit Monoclonal Antibody (102), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Invitrogen MA543356
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Specificity: Rat E-Selectin/CD62e/SELE
Leukocytes adhere to the blood vessel endothelium during extravasation in postcapillary venules of lymph nodes. In addition, leukocyte adhesion occurs in the capillaries and small venules at any site in the body after onset of inflammation. This response is immediate and involves a cascade of adhesion receptors. At the endothelial surface members of the selectin and immunoglobulin superfamilies participate in this cascade. Selectins are C-type cell surface lectins that play a role in leukocyte adhesion to the blood vessel wall endothelium. E-selectin (CD62E) is an endothelial cell specific selectin that is expressed only after activation with proinflammatory cytokines. In vitro experiments have shown that IL-1, TNFalpha and bacterial wall components like lipopolysaccharides induce the transcription of E-selectin in a NFkB dependent signaling cascade. E-selectin has been associated with blood vessel endothelium in diverse inflammatory situations.
Especificaciones
E-selectin | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P98105 | |
SELE | |
Recombinant Rat E-Selectin/CD62e/SELE Protein. | |
50 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
ELISA | |
102 | |
Unconjugated | |
SELE | |
CD62 antigen-like family member E; CD62E; e selectin; Elam; ELAM1; ELAM-1; endothelial adhesion molecule 1; endothelial leukocyte adhesion molecule; endothelial leukocyte adhesion molecule 1; endothelial leukocyte adhesion molecule I; ESEL; E-selectin; LECAM2; leukocyte endothelial cell adhesion molecule 2; leukocyte-endothelial cell adhesion molecule 2; RP1-117P20.2; SELE; selectin E; selectin E (endothelial adhesion molecule 1); selectin, endothelial cell | |
Rabbit | |
Protein A | |
RUO | |
25544 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
E-selectin Recombinant Rabbit Monoclonal Antibody (102), Invitrogen™ > 50 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido