missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Dynamin 1 Monoclonal Antibody (A1B2)
Mouse Monoclonal Antibody
Marca: Invitrogen™ MA534956
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Positive Control: PC-3M cell lysates, rat brain tissue, mouse brain tissue, A549.
Dynamin is a GTPase required for endocytosis of clathrin coated vesicles from the membrane. Dynamin's involvement in endocytosis was first discovered during the characterization of the temperature sensitive parylytic shibire mutation from Drosophila melanogaster. Three closely related isoforms have been identified in mammals. Dynamin1 is expressed exclusively in neurons, dynamin2 is ubiquitously expressed and dynamin3 is expressed primarily in the testes. Each of these isoforms of dynamin also has several splice variants. This suggests that each isoform/splice variant may have separate intracellular targets or function in the budding of distinct vesicle types. Common domain features of the dynamin proteins include an amino acid terminal GTPase domain, a central pleckstrin homology domain and a highly basic proline rich carboxyl terminus with several Src homology 3 (SH3) domains. These SH3 domains target the dynamin to the clathrin coated pit. Dynamin is known to interact with at least two proteins, AP2 adaptor protein complex and amphiphysin.
Especificaciones
Dynamin 1 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
Q05193 | |
DNM1 | |
Recombinant protein within Human Dynamin 1 aa 555-770. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
A1B2 | |
Unconjugated | |
DNM1 | |
AI838169; B-dynamin; D100; Dnm; Dnm1; Dyn1; dynamin 1; Dynamin BREDNM19; dynamin i; dynamin, brain; Dynamin1; Dynamin-1; EC 3.6.5.5; EIEE31; Ftfl; Kiaa4093; mKIAA4093; Neuronal dynamin; OTTHUMP00000215500 | |
Mouse | |
Protein A | |
RUO | |
1759 | |
-20°C, Avoid Freeze/Thaw Cycles, store in dark | |
Liquid |
Sugerencias de productos
Los clientes que vieron este artículo también vieron
Viewing 1-4 of
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Invitrogen™ Dynamin 1 Monoclonal Antibody (A1B2) > 100 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido