missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dopamine D1R/DRD1 Antibody (SG2-D1a), Novus Biologicals™
Mouse Monoclonal Antibody
172.00€ - 520.00€
Especificaciones
Antígeno | Dopamine D1R/DRD1 |
---|---|
Clon | SG2-D1a |
Concentración | 1.0 mg/mL |
Dilución | Western Blot 1:200-1:500, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 31462765), Immunohistochemistry-Frozen 1:1000, Immunohistochemistry Free-Floating |
Aplicaciones | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Free Floating) |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
18294380
|
Bio-Techne
NB110-60017SS |
0.025 mL | N/A | ||||||
18758793
|
Novus Biologicals
NB110-60017 |
0.1 mL |
520.00€
0.10 ml |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
Dopamine D1R/DRD1 Monoclonal specifically detects Dopamine D1R/DRD1 in Mouse, Rat, Bovine (Negative), Canine (Negative), Human (Negative), Porcine (Negative), Rabbit (Negative), Sheep (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating.Especificaciones
Dopamine D1R/DRD1 | |
1.0 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Free Floating) | |
Unconjugated | |
Mouse | |
GPCR, Neuroscience, Neurotransmission | |
D(1A) dopamine receptor, D1DR, DADR, Dopamine D1 receptor, dopamine receptor D1, DRD1A | |
DRD1 | |
IgG2b κ | |
Protein A purified | |
This antibody is specific for Dopamine Receptor D1A. There is no cross-reactivity with D1B (D5) receptor. |
SG2-D1a | |
Western Blot 1:200-1:500, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 31462765), Immunohistochemistry-Frozen 1:1000, Immunohistochemistry Free-Floating | |
Monoclonal | |
Purified | |
RUO | |
Mouse, Rat, Bovine (Negative), Canine (Negative), Human (Negative), Porcine (Negative), Rabbit (Negative) | |
1812 | |
Recombinant rat Dopamine Receptor D1 protein near the C-terminus. [Swiss-Prot# P18901] | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Dopamine D1R/DRD1 Antibody (SG2-D1a), Novus Biologicals™