missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dopamine D1R/DRD1 Antibody (SG2-D1a), Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Bio-Techne NB110-60017SS
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Dopamine D1R/DRD1 Monoclonal specifically detects Dopamine D1R/DRD1 in Mouse, Rat, Bovine (Negative), Canine (Negative), Human (Negative), Porcine (Negative), Rabbit (Negative), Sheep (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating.
Especificaciones
Dopamine D1R/DRD1 | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1:200-1:500, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 31462765), Immunohistochemistry-Frozen 1:1000, Immunohistochemistry Free-Floating | |
DRD1 | |
Recombinant rat Dopamine Receptor D1 protein near the C-terminus. [Swiss-Prot# P18901] | |
0.025 mL | |
GPCR, Neuroscience, Neurotransmission | |
1812 | |
Mouse, Rat, Bovine (Negative), Canine (Negative), Human (Negative), Porcine (Negative), Rabbit (Negative) | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Free Floating) | |
SG2-D1a | |
Unconjugated | |
D(1A) dopamine receptor, D1DR, DADR, Dopamine D1 receptor, dopamine receptor D1, DRD1A | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
This antibody is specific for Dopamine Receptor D1A. There is no cross-reactivity with D1B (D5) receptor. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG2b κ |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Dopamine D1R/DRD1 Antibody (SG2-D1a), Novus Biologicals™ > 0.025mL; Unlabeled
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido