Learn More
Dextran Mouse, FITC, Clone: DX1, Isotype Control, STEMCELL Technologies
Mouse Monoclonal Antibody

159.30€
Especificaciones
Antígeno | Dextran |
---|---|
Clon | DX1 |
Aplicaciones | Functional Assay, ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
Clasificación | Monoclonal |
Conjugado | FITC |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
17168581
![]() |
Stemcell Technologies
60026FI.1 |
25 Tests |
159.30€
25 tests |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
- The DX1 antibody reacts with dextran, a natural polysaccharide consisting of linear chains of alpha-1,6-linked D-glucopyranose residues with short side chains (mostly 1 - 2 glucose units) 1,3- or 1,4-linked to the backbone of the biopolymer
- Dextran has several uses as an additive in food, in lacquers, as a plasma volume expander, and as a coating for particles used in bioimaging or cell separation applications
- The DX1 antibody was raised against isomaltotetraose (four sugar units in length) but binds better to longer polymer chains
- This antibody can be used to detect cells labelled with dextran-coated nanoparticles during StemSep™ and EasySep™ cell separation procedures or for other applications that require the detection of dextran
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Human T Cell Enrichment Kit (Catalog #19051) and EasySep™ Human CD4+ T Cell Enrichment Kit (Catalog #19052)
Especificaciones
Dextran | |
Functional Assay, ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
FITC | |
IgG1 κ |
DX1 | |
Monoclonal | |
Mouse | |
Isotype Control (Primary) |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.