Learn More
ACROBiosystems Cynomolgus / Rhesus macaque FOLR1 Protein, His tag
Marca: ACROBiosystems FO1-C52H8-1mg

Descripción
Folate Receptor 1 (FOLR1) is also known as Folate receptor alpha, Folate Binding Protein (FBP), FOLR, and is a member of the folate receptor (FOLR) family. Members of this gene family have a high affinity for folic acid and for several reduced folic acid derivatives, and mediate delivery of 5-methyltetrahydrofolate to the interior of cells. Mature FOLR1 is an N-glycosylated protein that is anchored to the cell surface by a GPI linkage. FOLR1 is predominantly expressed on epithelial cells and is dramatically upregulated on many carcinomas. FOLR1 is internalized to the endosomal system where it dissociates from its ligand before recycling to the cell surface. A soluble form of FOLR1 can be proteolytically shed from the cell surface into the serum and breast milk. Defects in FOLR1 are the cause of neurodegeneration due to cerebral folate transport deficiency (NCFTD). NCFTD is an autosomal recessive disorder resulting from brain-specific folate deficiency early in life.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.4 kDa. The protein migrates as 32-41 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_001181576.1 | |
1 mg | |
Arg 25 - Met 233 | |
FOLR-1,FBP,FOLR | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
26.4 kDa | |
Cynomolgus / Rhesus macaque FOLR1, His Tag (FO1-C52H8) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (Accession # NP_001181576.1). In the region Arg 25 - Met 233, the AA sequence of Cynomolgus and Rhesus macaque FOLR1 are homologus. | |
-20°C | |
FOLR1 | |
Cynomolgus Monkey, Rhesus Macaque | |
Tris and Glycine, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.