Learn More
Invitrogen™ Human CSF1R (FMS), His Tag Recombinant Protein
Recombinant Protein
Marca: Invitrogen™ PV4092
Descripción
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Cytoplasmic
CSF1R (FMS) is involved in the production, differentiation, and function of macrophages, and is implicated in promyelocytic leukemias. Ligand binding activates the receptor kinase through a process of oligomerization and transphosphorylation. CSF1R is a tyrosine kinase transmembrane receptor and member of the CSF1/PDGF receptor family of tyrosine-protein kinases. Mutations in the gene encoding CSF1R have been associated with a predisposition to myeloid malignancy.Tyrosine-protein kinase that acts as cell-surface receptor for CSF1 and IL34 and plays an essential role in the regulation of survival, proliferation and differentiation of hematopoietic precursor cells, especially mononuclear phagocytes, such as macrophages and monocytes. Promotes the release of proinflammatory chemokines in response to IL34 and CSF1, and plays an important role in innate immunity and in inflammatory processes. The first intron of the CSF1R gene contains a transcriptionally inactive ribosomal protein L7 processed pseudogene oriented in the opposite direction. Mutations in the CSF1R gene have been associated with a predisposition to myeloid malignancy.
Especificaciones
P07333 | |
Kinase Assay | |
1436 | |
Human CSF1R (FMS), His Tag | |
AI323359; CD115; CD115 antigen; C-FMS; colony stimulating factor 1 receptor; colony stimulating factor 1 receptor, formerly McDonough feline sarcoma viral (v-fms) oncogene homolog; CSF-1 receptor; CSF1R; CSF-1 R; CSF-1-R; CSFIR; Csfmr; CSFR; EC 2.7.10.1; FIM2; Fim-2; FMS; fms proto-oncogene; HDLS; kinase CSFR; macrophage colony stimulating factor I receptor; macrophage colony-stimulating factor 1 receptor; McDonough feline sarcoma viral (v-fms) oncogene homolog; M-CSFR; M-CSF-R; mrfms; OTTHUMP00000224093; protein-tyrosine kinase; proto-oncogene c-Fms; proto-oncogene fms | |
PDGFR Kinase Family | |
Unconjugated | |
Recombinant | |
-80°C, Avoid Freeze/Thaw Cycles | |
Dry Ice | |
Cytoplasmic | |
Drug Development |
See Label | |
50 mM tris with 0.05% Triton X-100, 0.05 mM EDTA, 100 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5 | |
44.8 kDa | |
1 mg | |
CSF1R (CD115) | |
538-910 | |
Human | |
His-tag | |
Tyrosine Kinases | |
Baculovirus | |
Liquid |
Certificados
Se requiere un número de lote para mostrar resultados para certificados. Para encontrar su número de lote en pedidos anteriores, utilice el área de estado de pedidos.
Número de lote | Tipo de certificado | Fecha | Catalog Number |
---|---|---|---|
2475673D | Certificado de análisis | 08/08/2022 | PV4092 |
2112355J | Certificado de análisis | 19/01/2022 | PV4092 |
1883684E | Certificado de análisis | 15/11/2017 | PV4092 |
1883684C | Certificado de análisis | 19/07/2017 | PV4092 |
662393M | Certificado de análisis | 14/04/2017 | PV4092 |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.