missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRIP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15349-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CRIP1 is a cysteine-rich intestinal protein (CRIP) that belongs to the LIM/double zinc finger protein family, members of which include cysteine- and glycine-rich protein-1 (CSRP1 MIM 123876), rhombotin-1 (RBTN1 MIM 186921), rhombotin-2 (RBTN2 MIM 180385), and rhombotin-3 (RBTN3 MIM 180386). CRIP may be involved in intestinal zinc transport.Especificaciones
CRIP1 | |
Polyclonal | |
Unconjugated | |
CRIP1 | |
CRHP, CRIP, CRIP1, CRP 1, CRP1, Cysteine rich heart protein, Cysteine rich protein 1, hCRHP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1396 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P50238 | |
CRIP1 | |
CRIP1 Fusion Protein Ag7588 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
CRIP1 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur