missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cre Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NB100-56134
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Cre Polyclonal antibody specifically detects Cre in Mouse, Cre/lox System samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), SDS-Page, Immunohistochemistry Free-Floating
Especificaciones
Cre | |
Polyclonal | |
Western Blot 1:1000 - 1:2000. Use reported in scientific literature (PMID 33784255), Immunohistochemistry 1:10 - 1:5000, Immunoprecipitation 1:50 - 1:200, Immunohistochemistry-Paraffin 1:1000 - 1:5000, Immunohistochemistry-Frozen 1:10 - 1:5000, SDS-Page reported in scientific literature (PMID 27432829), Immunohistochemistry Free-Floating reported in scientific literature (PMID 25429143) | |
Cyclization recombinase | |
A synthethic peptide corresponding to a portion of amino acids 1-50 of the bacteriophage protein CRE was used as immunogen, GenBank no. AAQ14086.1. | |
0.05 mL | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
50 uL neat whole antisera | |
Rabbit | |
Unpurified | |
RUO | |
Mouse, Cre/lox System | |
Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Cre Antibody, Novus Biologicals™ > 0.05 mL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido