missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated NQO1 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech CL488-11451-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The NQO1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NQO1. This antibody recognizes human antigen. The NQO1 antibody has been validated for the following applications: IF/ICC, FC (Intra) analysis. This antibody is coralite™ plus 488.Especificaciones
NQO1 | |
Polyclonal | |
CoraLite Plus 488 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
1728 | |
Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
Conjugated Antibody | |
IgG |
Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
1000 μg/mL | |
DHQU; DIA4; DTD; NMOR1; NMORI; QR1 | |
Recombinant Protein | |
20 μL | |
Primary | |
Human | |
31 kDa | |
Liquid |
Sugerencias de productos
Customers who viewed this item also viewed.
Viewing 1-4 of
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
CoraLite™ Plus 488-conjugated NQO1 Rabbit Polyclonal Antibody, Proteintech > CoraLite™ Plus 488; 20 μL
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido