Learn More
Fisherbrand™ Tapones de celulosa
Para matraces, erlenmeyer y tubos de ensayo.
Brand:  Fisherbrand™ 0005N00053
 View more versions of this product
Product Code. 11768513
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
View alternative products »
This item is not returnable. View return policy
Description
Description
  • Permeable en el aire
  • Esterilizable a una temperatura máxima de 200 °C
  • Un solo uso
Specifications
Specifications
30 mm
200 °C
7 mm
Tapón
Vasos de precipitados, matraces Erlenmeyer, tubos y diferentes tipos de frascos
Paq. de 1000
7 mm
Fibra de celulosa
Estériles
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Tapones de celulosa > Size: 7 x 11 x 30mm
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.