Learn More
Fisherbrand™ Tapones de celulosa
Para matraces, erlenmeyer y tubos de ensayo.
Marca:  Fisherbrand™ 0005N00057
 Ver más versiones de este producto
Código de producto. 11740096
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos »
Descripción
Descripción
  • Permeable en el aire
  • Esterilizable a una temperatura máxima de 200 °C
  • Un solo uso
Especificaciones
Especificaciones
32 mm
200 °C
16 mm
Stopper
Vasos de precipitados, matraces Erlenmeyer, tubos y diferentes tipos de frascos
1000PK
11 mm
Cellulose Fiber
Estériles
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Tapones de celulosa > Size: 11 x 16 x 32mm
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.