missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD7 Recombinant Rabbit Monoclonal Antibody (15), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Invitrogen MA543531
Detalles adicionales : Peso : 0.01000kg
Descripción
Specificity: Human CD7/LEU9
CD7 Recombinant Monoclonal antibody specifically detects CD7 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Especificaciones
CD7 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P09564 | |
CD7 | |
Recombinant Human CD7 protein. | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), ELISA | |
15 | |
Unconjugated | |
CD7 | |
Cd7; CD7 antigen; CD7 antigen (p41); Cd7 molecule; GP40; LEU-9; p41 protein; T-cell antigen CD7; T-cell leukemia antigen; T-cell surface antigen Leu-9; Tp40; TP41 | |
Rabbit | |
Protein A | |
RUO | |
924 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |