missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD62L Recombinant Rabbit Monoclonal Antibody (018), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Invitrogen MA543355
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Specificity: Human CD62L/L-Selectin/SELL
The human CD62L is a 74-95 kDa glycoprotein member of the selectin family of adhesion receptors. L-Selectin is comprised of an aminoterminal C-type lectin binding domain, an epidermal growth factor-like domain, two short consensus repeat (SCR) sequences homologous to those found in complement binding proteins, a short spacer region, a transmembrane region and a short cytoplasmic region. Human CD62L (L-Selectin) is constitutively expressed on all classes of leukocytes including lymphocytes (except a substantial population of memory T cells), monocytes and polymorphonuclear cells.
Especificaciones
CD62L | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P14151 | |
SELL | |
Recombinant Human CD62L/L-Selectin/SELL protein. | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA | |
018 | |
Unconjugated | |
SELL | |
A.11; AI528707; CD62 antigen-like family member L; CD62L; gp90-MEL; hLHRc; LAM1; LAM-1; LECAM1; LECAM-1; LEU8; Leu-8; Leukocyte adhesion molecule 1; leukocyte surface antigen Leu-8; Leukocyte-endothelial cell adhesion molecule 1; LNHR; LSEL; L-selectin; Ly22; Ly-22; LYAM1; Lyam-1; Ly-m22; Lymph node homing receptor; lymphocyte adhesion molecule 1; lymphocyte antigen 22; Lymphocyte surface MEL-14 antigen; pln homing receptor; PLNHR; SELE; selectin E (endothelial adhesion molecule 1); selectin L; selectin L (lymphocyte adhesion molecule 1); selectin, lymphocyte; Sell; TQ1 | |
Rabbit | |
Protein A | |
RUO | |
6402 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
CD62L Recombinant Rabbit Monoclonal Antibody (018), Invitrogen™ > 50 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido