Learn More
CD3 Molecular Complex Rat anti-Mouse, PE, Clone: 17A2, BD
Rat Monoclonal Antibody
Marca: BD Biosciences 555275
395.22 EUR hasta el 2025-12-31
Use el código promocional "25339" para aplicar la oferta.
Descripción
The 17A2 monoclonal antibody specifically binds to the T-cell receptor-associated CD3 complex that is expressed on many thymocytes and mature T lymphocytes. Plate-bound 17A2 antibody has been reported to induce IL-2 production by cultured T cells in the absence of accessory cells. The binding of the 17A2 antibody to T cells can be blocked by the anti-CD3e mAb 145-2C11. This suggests that the 17A2 antibody recognizes an epitope of the CD3 epsilon chain. In vivo treatment with 17A2 antibody has been reported to partially deplete T lymphocytes and temporarily down-modulates CD3 expression on T cells.

Especificaciones
CD3 Molecular Complex | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
γδ TCR-positive T-T hybridoma D1 | |
0.2 mg | |
Immunology | |
Mouse | |
IgG2b κ |
Flow Cytometry | |
17A2 | |
PE | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only.