missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD223 (LAG-3) Monoclonal Antibody (3DS223H), NovaFluor™ Yellow 610, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen H048T03Y03
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
LAG-3 is a 70-kDa surface glycoprotein belonging to the Ig superfamily with homology to CD4. LAG-3 binds to MHC class II with higher affinity than CD4 and is thought to be involved in the negative regulation of T cell activation and homeostatic proliferation. Surface expression of LAG-3 has been reported on activated T cells (including regulatory T cells) and NK cells. CD8+ T cells usually express LAG-3 at significantly higher levels than CD4+ T cells. Coexpression of LAG-3 and CD49b has been proposed to identify human and mouse Type 1 regulatory T cells (Tr1 cells).
Especificaciones
CD223 (LAG-3) | |
Monoclonal | |
0.4 μg/Test | |
PBS with 0.09% sodium azide; pH 7.2 | |
P18627 | |
LAG3 | |
Affinity chromatography | |
RUO | |
3902 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
3DS223H | |
NovaFluor Yellow 610 | |
LAG3 | |
Activation-induced cytidine deaminase-linked autoimmunity protein; Aida; CD223; FDC; LAG3; LAG-3; Ly66; lymphocyte activating 3; lymphocyte activation gene 3 protein; lymphocyte-activation gene 3; Secreted lymphocyte activation gene 3 protein; sLAG 3; sLAG3; sLAG-3; soluble LAG 3lymphocyte activating 3; soluble LAG3 | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
CD223 (LAG-3) Monoclonal Antibody (3DS223H), NovaFluor™ Yellow 610, eBioscience™, Invitrogen™ > 100 Tests; NovaFluor Yellow 610
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido