missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD146 Rat anti-Mouse, BUV563, Clone: ME-9F1, BD Biosciences™
Rat Monoclonal Antibody
Marca: BD Biosciences 741443
441.74 EUR hasta el 2025-12-31
Use el código promocional "25339" para aplicar la oferta.
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CD146 Monoclonal antibody specifically detects CD146 in Mouse samples. It is validated for Flow Cytometry
Especificaciones
CD146 | |
Monoclonal | |
Brilliant Ultraviolet 563 | |
S-endo 1; S-endo; Muc18; Mcam; Gicerin | |
CBA/J Mouse lymph node Cell Line | |
RUO | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
ME-9F1 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV563 under optimal conditions that minimize unconjugated dye and antibody. |
Sugerencias de productos
Customers who viewed this item also viewed.
Viewing 1-5 of 12
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
CD146 Rat anti-Mouse, BUV563, Clone: ME-9F1, BD Biosciences™ > 50 μg; BUV563
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido