missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD123 Recombinant Rabbit Monoclonal Antibody (001), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Invitrogen MA543117
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Specificity: Mouse IL3Ra
IL-3Ra (Interleukin-3 receptor subunit alpha) belongs to the type I cytokine receptor family and is a heterodimer with a unique alpha chain paired with the common beta (beta c or CDw131) subunit. IL-3Ra is responsible for the transmission of signal of Interleukin-3. IL-3Ra and IL-3Rb chain heterodimerization is induced by Interleukin-3, which is required for receptor activation but not high affinity binding. IL-3Ra is also reported that IL-3Ra subunits are overexpressed on acute myeloid leukemia (AML) blasts compared with normal hematopoietic cells and are thus potential targets for novel therapeutic agents.
Especificaciones
CD123 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P26952 | |
IL3RA | |
Recombinant Mouse IL3Ra protein. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
ELISA | |
001 | |
Unconjugated | |
IL3RA | |
CD123; CD123 antigen; CDw123; Cyrl; cytokine receptor-like protein CYRL; hIL-3Ra; Il-3 alpha subunit; IL-3 receptor alpha chain; IL-3 receptor alpha SP2 isoform; IL-3 receptor subunit alpha; IL3R; IL-3R subunit alpha; Il3ra; IL-3RA; IL-3R-alpha; IL3RAY; IL3RX; IL3RY; interleukin 3 receptor subunit alpha; interleukin 3 receptor, alpha; interleukin 3 receptor, alpha (low affinity); interleukin 3 receptor, alpha chain; interleukin-3 receptor class II alpha chain; Interleukin-3 receptor subunit alpha; Sut-1 | |
Rabbit | |
Protein A | |
RUO | |
16188 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
CD123 Recombinant Rabbit Monoclonal Antibody (001), Invitrogen™ > 100 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido